Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
suz12a
Entrez Gene ID
794171
Gene Name
SUZ12 polycomb repressive complex 2 subunit a
Gene Aliases
suz12, suz12l, zgc:100863
Chromosome Location
Chr.3: 35849672 - 35865795 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003529.2 NP_001003529.1 14 - 15 1910 104
XM_021471664.1 XP_021327339.1 14 - 15 2922 104
GenBank mRNA BC078293.1 - 14 - 15 1913 104
BC152625.1 - 14 - 15 1799 104

View Full Product Details