Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thoc3
Entrez Gene ID
445302
Gene Name
THO complex 3
Gene Aliases
zgc:100815
Chromosome Location
Chr.14: 25505474 - 25510955 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003758.1 NP_001003758.1 4 - 5 716 88
GenBank mRNA BC079495.1 - 4 - 5 716 88
BC152622.1 - 4 - 5 710 88

View Full Product Details