Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cby1
Entrez Gene ID
326875
Gene Name
chibby homolog 1 (Drosophila)
Gene Aliases
LRP, fe37g01, wu:fe37g01
Chromosome Location
Chr.12: 424944 - 426514 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003838.1 NP_001003838.1 2 - 3 283 81
GenBank mRNA AF393213.1 - 2 - 3 283 81
BC171621.1 - 2 - 3 252 81

View Full Product Details