Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan13b
Entrez Gene ID
449797
Gene Name
tetraspanin 13b
Gene Aliases
tspan13, zgc:101833
Chromosome Location
Chr.19: 30387996 - 30399203 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005970.1 NP_001005970.1 4 - 5 614 63
GenBank mRNA BC083295.1 - 4 - 5 614 63
BC165008.1 - 4 - 5 458 63

View Full Product Details