Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dck
Entrez Gene ID
474325
Gene Name
deoxycytidine kinase
Gene Aliases
wu:fc15f06, zgc:101771
Chromosome Location
Chr.21: 5974676 - 5986172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007056.1 NP_001007057.1 4 - 5 592 68
GenBank mRNA BC083277.1 - 4 - 5 592 68
BC164599.1 - 4 - 5 584 68

View Full Product Details