Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdxka
Entrez Gene ID
791775
Gene Name
pyridoxal (pyridoxine, vitamin B6) kinase a
Gene Aliases
cb1056, fb10c12, pdxkl, si:dkey-281i8.2, wu:fa55a03, wu:fb10c12, zgc:101900
Chromosome Location
Chr.9: 19623406 - 19657400 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126449.1 NP_001119921.1 1 - 2 320 99
XM_021478775.1 XP_021334450.1 1 - 2 312 99
GenBank mRNA AI384995.1 - 1 - 2 251 99
BC085468.1 - 1 - 2 277 99
BC153430.1 - 1 - 2 274 99
BC164257.1 - 1 - 2 120 99
CF943692.1 - 1 - 2 265 99

View Full Product Details