Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftr72
Entrez Gene ID
492515
Gene Name
finTRIM family, member 72
Gene Aliases
zgc:103602
Chromosome Location
Chr.15: 41752335 - 41756507 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007387.2 NP_001007388.2 6 - 7 1200 150
GenBank mRNA BC085564.1 - 6 - 7 1200 150

View Full Product Details