Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pus1
Entrez Gene ID
494060
Gene Name
pseudouridine synthase 1
Gene Aliases
wu:fb98h11, zgc:103516
Chromosome Location
Chr.21: 14826584 - 14832369 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC086827.1 - 5 - 6 1303 78
EH559949.1 - 1 - 2 156 78

View Full Product Details