Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctsk
Entrez Gene ID
550475
Gene Name
cathepsin K
Gene Aliases
wu:fa95f03, wu:fb08b05, zgc:110367
Chromosome Location
Chr.16: 29492994 - 29502549 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017778.1 NP_001017778.1 6 - 7 873 77
GenBank mRNA BC092901.1 - 6 - 7 873 77
BC164579.1 - 5 - 6 829 77
HM239643.1 - 5 - 6 717 77
HM239644.1 - 5 - 6 717 77

View Full Product Details