Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mettl2a
Entrez Gene ID
100006618
Gene Name
methyltransferase like 2A
Gene Aliases
zgc:110598
Chromosome Location
Chr.3: 140550 - 153752 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017902.1 NP_001017902.1 8 - 9 927 99
GenBank mRNA BC092943.1 - 8 - 9 927 99
BC151872.1 - 7 - 8 939 99
BC154164.1 - 8 - 9 99

View Full Product Details