Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem41aa
Entrez Gene ID
553803
Gene Name
transmembrane protein 41aa
Gene Aliases
tmem41a, zgc:112259
Chromosome Location
Chr.9: 12444494 - 12455521 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020765.1 NP_001018601.1 1 - 2 234 78
GenBank mRNA BC095709.1 - 1 - 2 234 78

View Full Product Details