Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmem41aa
Entrez Gene ID
553803
Gene Name
transmembrane protein 41aa
Gene Aliases
tmem41a, zgc:112259
Chromosome Location
Chr.9: 12444494 - 12455521 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020765.1 NP_001018601.1 4 - 5 675 118
GenBank mRNA BC095709.1 - 4 - 5 675 118

View Full Product Details