Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hmbsb
Entrez Gene ID
338290
Gene Name
hydroxymethylbilane synthase, b
Gene Aliases
PBGD, hmbsl, id:ibd5004, im:7140060, zgc:110690
Chromosome Location
Chr.15: 23511403 - 23523222 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024388.1 NP_001019559.1 5 - 6 389 88
XM_005157569.4 XP_005157626.1 5 - 6 372 88
GenBank mRNA AB188810.1 - 5 - 6 347 88
BC095373.1 - 5 - 6 389 88

View Full Product Details