Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
camk2g1
Entrez Gene ID
791855
Gene Name
calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma 1
Gene Aliases
camk2ga, zgc:109908
Chromosome Location
Chr.12: 35345824 - 35505623 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030125.1 NP_001025296.1 19 - 20 1523 76
XM_005156361.4 XP_005156418.1 21 - 22 1771 76
XM_005156362.4 XP_005156419.1 21 - 22 1762 76
XM_005156363.4 XP_005156420.1 20 - 21 1708 76
XM_005156364.4 XP_005156421.1 20 - 21 1685 76
XM_005156365.4 XP_005156422.1 20 - 21 1632 76
XM_005156366.4 XP_005156423.1 19 - 20 1612 76
XM_005156367.4 XP_005156424.1 19 - 20 1527 76
XM_005156368.4 XP_005156425.1 19 - 20 1517 76
XM_005156369.4 XP_005156426.1 18 - 19 1454 76
XM_005156370.4 XP_005156427.1 18 - 19 1431 76
XM_005156371.4 XP_005156428.1 17 - 18 1367 76
XM_009306817.3 XP_009305092.1 19 - 20 1621 76
XM_021480164.1 XP_021335839.1 20 - 21 1675 76
XM_021480165.1 XP_021335840.1 20 - 21 1622 76
XM_021480166.1 XP_021335841.1 18 - 19 1440 76
GenBank mRNA BC096785.1 - 19 - 20 1523 76

View Full Product Details