Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab8b
Entrez Gene ID
100093706
Gene Name
RAB8B, member RAS oncogene family
Gene Aliases
rab8bb, zgc:165529
Chromosome Location
Chr.7: 186105 - 193970 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099259.1 NP_001092729.1 5 - 6 519 91
GenBank mRNA BC142846.1 - 5 - 6 519 91

View Full Product Details