Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kri1
Entrez Gene ID
402800
Gene Name
KRI1 homolog
Gene Aliases
flj12949l, kri1l
Chromosome Location
Chr.3: 19336501 - 19355200 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002041.1 NP_001002041.1 3 - 4 359 141
GenBank mRNA AY648765.1 - 3 - 4 359 141
BC045326.1 - 3 - 4 360 141
BC163908.1 - 3 - 4 348 141

View Full Product Details