Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aqp10a
Entrez Gene ID
436621
Gene Name
aquaporin 10a
Gene Aliases
aqp10, zgc:92167
Chromosome Location
Chr.16: 52637 - 59827 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002349.1 NP_001002349.1 3 - 4 422 67
GenBank mRNA BC075911.1 - 3 - 4 422 67

View Full Product Details