Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
galm
Entrez Gene ID
436646
Gene Name
galactose mutarotase
Gene Aliases
zgc:92501
Chromosome Location
Chr.13: 8306407 - 8333891 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002373.1 NP_001002373.1 1 - 2 247 76
XM_021480558.1 XP_021336233.1 1 - 2 288 76
GenBank mRNA BC076039.1 - 1 - 2 247 76
BC153585.1 - 1 - 2 240 76

View Full Product Details