Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psmd9
Entrez Gene ID
436709
Gene Name
proteasome 26S subunit, non-ATPase 9
Gene Aliases
wu:fb30g07, zgc:92643
Chromosome Location
Chr.7: 72003301 - 72011167 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002436.2 NP_001002436.2 3 - 4 609 77
XM_005169066.4 XP_005169123.1 3 - 4 595 77
GenBank mRNA AI437369.1 - 3 - 4 77
BC076129.1 - 3 - 4 575 77
BC096994.1 - 3 - 4 601 77

View Full Product Details