Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nudt13
Entrez Gene ID
556815
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 13
Gene Aliases
si:ch211-146f4.4, zC146F4.4
Chromosome Location
Chr.12: 253145 - 267851 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044816.1 NP_001038281.1 2 - 3 248 108
XM_009306499.2 XP_009304774.1 4 - 5 1053 108

View Full Product Details