Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
amn1
Entrez Gene ID
751744
Gene Name
antagonist of mitotic exit network 1 homolog (S. cerevisiae)
Gene Aliases
zgc:153121
Chromosome Location
Chr.25: 28893631 - 28903584 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045454.1 NP_001038919.1 3 - 4 435 85
GenBank mRNA BC122149.1 - 3 - 4 435 85
BC154132.1 - 3 - 4 435 85

View Full Product Details