Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fam98b
Entrez Gene ID
780841
Gene Name
family with sequence similarity 98 member B
Gene Aliases
zgc:158382
Chromosome Location
Chr.17: 220719 - 225527 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079662.1 NP_001073130.1 1 - 2 126 81
XM_017351682.2 XP_017207171.1 1 - 2 195 81
GenBank mRNA BC127591.1 - 1 - 2 126 81
BC153522.1 - 1 - 2 109 81

View Full Product Details