Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rrp36
Entrez Gene ID
321183
Gene Name
ribosomal RNA processing 36
Gene Aliases
cb268, sb:cb268, si:dkey-236e20.6
Chromosome Location
Chr.22: 15343953 - 15352546 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079944.1 NP_001073413.1 6 - 7 951 124
GenBank mRNA BC142737.1 - 6 - 7 949 124

View Full Product Details