Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kdm2aa
Entrez Gene ID
571851
Gene Name
lysine (K)-specific demethylase 2Aa
Gene Aliases
jhdm1A-b, wu:fb76b11, wu:fj11c04, zgc:158441, zgc:158606
Chromosome Location
Chr.1: 44838706 - 44869972 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081692.2 NP_001075161.2 20 - 21 3692 70
XM_005160248.4 XP_005160305.1 20 - 21 3650 70
XM_009293357.3 XP_009291632.1 20 - 21 4278 70
XM_009293362.3 XP_009291637.1 19 - 20 3490 70
XM_009293365.3 XP_009291640.1 20 - 21 3579 70
GenBank mRNA BC128874.1 - 11 - 12 2497 70

View Full Product Details