Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
surf1
Entrez Gene ID
557970
Gene Name
SURF1 cytochrome c oxidase assembly factor
Gene Aliases
im:6898613, zgc:158646
Chromosome Location
Chr.5: 33281208 - 33283718 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080173.1 NP_001073642.1 2 - 3 170 80
GenBank mRNA BC129348.1 - 2 - 3 170 80
BC164262.1 - 2 - 3 174 80

View Full Product Details