Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pomcb
Entrez Gene ID
100034412
Gene Name
proopiomelanocortin b
Gene Aliases
si:ch211-203k16.2
Chromosome Location
Chr.20: 42968607 - 42978321 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083051.1 NP_001076520.1 1 - 2 127 73
XM_005170169.4 XP_005170226.1 2 - 3 200 73

View Full Product Details