Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nos2b
Entrez Gene ID
563654
Gene Name
nitric oxide synthase 2b, inducible
Gene Aliases
inosb
Chromosome Location
Chr.15: 30323491 - 30348935 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113501.1 NP_001106973.1 13 - 14 1739 82
XM_005157694.4 XP_005157751.1 5 - 6 502 82
XM_009291655.3 XP_009289930.1 12 - 13 2796 82
XM_017350981.2 XP_017206470.1 12 - 13 2794 82
GenBank mRNA AM749802.1 - 13 - 14 1739 82
EU332350.1 - 13 - 14 1590 82

View Full Product Details