Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfb2m
Entrez Gene ID
569468
Gene Name
transcription factor B2, mitochondrial
Gene Aliases
sb:cb857
Chromosome Location
Chr.17: 12058468 - 12073356 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113617.2 NP_001107089.2 3 - 4 651 64
GenBank mRNA BC154809.1 - 3 - 4 610 64
BC171547.1 - 3 - 4 574 64
EH486004.1 - 3 - 4 643 64
EH580492.1 - 3 - 4 677 64

View Full Product Details