Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nkain4
Entrez Gene ID
100002876
Gene Name
sodium/potassium transporting ATPase interacting 4
Gene Aliases
zgc:172210
Chromosome Location
Chr.23: 14263683 - 14404128 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113646.1 NP_001107118.1 6 - 7 836 105
GenBank mRNA BC154589.1 - 6 - 7 836 105
BC171601.1 - 6 - 7 736 105

View Full Product Details