Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
mkxb
Entrez Gene ID
556167
Gene Name
mohawk homeobox b
Gene Aliases
-
Chromosome Location
Chr.2: 3986083 - 4009200 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114409.1 NP_001107881.1 2 - 3 383 65
GenBank mRNA EU280786.1 - 2 - 3 383 65

View Full Product Details