Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thbs4a
Entrez Gene ID
563429
Gene Name
thrombospondin 4a
Gene Aliases
zgc:175276
Chromosome Location
Chr.5: 51909582 - 51942771 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114424.2 NP_001107896.2 14 - 15 1992 103
GenBank mRNA BC157385.1 - 14 - 15 1951 103

View Full Product Details