Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thbs4a
Entrez Gene ID
563429
Gene Name
thrombospondin 4a
Gene Aliases
zgc:175276
Chromosome Location
Chr.5: 51909582 - 51942771 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114424.2 NP_001107896.2 20 - 21 2969 97
GenBank mRNA BC157385.1 - 20 - 21 2928 97

View Full Product Details