Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bfb
Entrez Gene ID
30489
Gene Name
complement component bfb
Gene Aliases
Bf-2
Chromosome Location
Chr.21: 27437934 - 27443999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131241.1 NP_571316.1 7 - 8 1015 139
XM_021468750.1 XP_021324425.1 7 - 8 1062 139
GenBank mRNA AF047412.1 - 5 - 6 713 139
BC076051.1 - 7 - 8 1015 139

View Full Product Details