Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rh50
Entrez Gene ID
791729
Gene Name
Rh50-like protein
Gene Aliases
Rhp2
Chromosome Location
Chr.8: 37259395 - 37263524 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126444.1 NP_001119916.1 1 - 2 465 63
NM_131547.1 NP_571622.1 1 - 2 465 63
GenBank mRNA AF209468.1 - 1 - 2 465 63

View Full Product Details