Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mafaa
Entrez Gene ID
100000492
Gene Name
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog Aa
Gene Aliases
SMaf1, mafa, mafl
Chromosome Location
Chr.6: 32699363 - 32703518 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082940.2 NP_001076409.1 1 - 1 1837 67
GenBank mRNA AB006324.2 - 1 - 1 1850 67
BC133072.1 - 1 - 1 1636 67
BC162337.1 - 1 - 1 1661 67
BC162611.1 - 1 - 1 1661 67
EH467419.1 - 1 - 1 200 67

View Full Product Details