Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klf3
Entrez Gene ID
117603
Gene Name
Kruppel-like factor 3 (basic)
Gene Aliases
klf12
Chromosome Location
Chr.1: 247853 - 269091 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131859.3 NP_571934.3 4 - 5 721 119
XM_017356404.2 XP_017211893.1 4 - 5 830 119
GenBank mRNA AF392993.1 - 4 - 5 728 119
BC066635.1 - 4 - 5 692 119
EH602292.1 - 3 - 4 811 119

View Full Product Details