Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
depdc1a
Entrez Gene ID
100003959
Gene Name
DEP domain containing 1a
Gene Aliases
cb41, depdc1, sb:cb41, wu:fc05g03, zgc:55374
Chromosome Location
Chr.8: 65829 - 78118 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198872.1 NP_942573.1 2 - 3 534 68
XM_005166773.3 XP_005166830.1 2 - 3 535 68
GenBank mRNA BC044382.1 - 2 - 3 534 68
BC171596.1 - 2 - 3 486 68
BC171624.1 - 2 - 3 486 68

View Full Product Details