Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
depdc1a
Entrez Gene ID
100003959
Gene Name
DEP domain containing 1a
Gene Aliases
cb41, depdc1, sb:cb41, wu:fc05g03, zgc:55374
Chromosome Location
Chr.8: 65829 - 78118 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198872.1 NP_942573.1 3 - 4 669 104
XM_005166773.3 XP_005166830.1 3 - 4 670 104
GenBank mRNA BC044382.1 - 3 - 4 669 104
BC171596.1 - 3 - 4 621 104
BC171624.1 - 3 - 4 621 104

View Full Product Details