Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sult2st1
Entrez Gene ID
338214
Gene Name
sulfotransferase family 2, cytosolic sulfotransferase 1
Gene Aliases
id:ibd1140, sult2st
Chromosome Location
Chr.12: 630295 - 645972 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198914.2 NP_944596.2 4 - 5 736 94
XM_005155998.3 XP_005156055.1 4 - 5 1048 94
GenBank mRNA AY181063.1 - 4 - 5 580 94
BC064294.1 - 4 - 5 736 94

View Full Product Details