Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cfap298
Entrez Gene ID
792532
Gene Name
cilia and flagella associated protein 298
Gene Aliases
c10h21orf59, c21orf59, c21orf59l, grb, kur, sb:cb615, zgc:64057
Chromosome Location
Chr.10: 126598 - 131874 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200088.1 NP_956382.1 5 - 6 628 103
GenBank mRNA AY648836.1 - 5 - 6 609 103
BC053230.1 - 5 - 6 628 103
BC154333.1 - 4 - 5 649 103
BC171713.1 - 5 - 6 613 103
BC171715.1 - 5 - 6 613 103

View Full Product Details