Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wdr45b
Entrez Gene ID
393210
Gene Name
WD repeat domain 45B
Gene Aliases
WIPI-3, wdr45l, zgc:56002
Chromosome Location
Chr.12: 40351 - 46985 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200240.2 NP_956534.2 9 - 10 1368 81
GenBank mRNA BC047802.1 - 9 - 10 1024 81
BC154337.1 - 9 - 10 1015 81
BC164151.1 - 9 - 10 957 81

View Full Product Details