Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
selenbp1
Entrez Gene ID
393542
Gene Name
selenium binding protein 1
Gene Aliases
MTO, zgc:65844
Chromosome Location
Chr.16: 112115 - 135091 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200570.1 NP_956864.1 5 - 6 656 52
XM_005158207.2 XP_005158264.1 5 - 6 720 52
GenBank mRNA BC056590.1 - 5 - 6 656 52
MG797500.1 - 4 - 5 481 52

View Full Product Details