Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
timm17b
Entrez Gene ID
562144
Gene Name
translocase of inner mitochondrial membrane 17 homolog B (yeast)
Gene Aliases
si:ch211-163b2.6
Chromosome Location
Chr.8: 44921123 - 44926077 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113593.1 NP_001107065.1 4 - 5 338 65

View Full Product Details