Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mxg
Entrez Gene ID
360148
Gene Name
myxovirus (influenza virus) resistance G
Gene Aliases
-
Chromosome Location
Chr.25: 19695429 - 19715137 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122971.1 NP_001116443.1 10 - 11 1420 86
XM_009297890.2 XP_009296165.1 10 - 11 1643 86
XM_021470456.1 XP_021326131.1 8 - 9 1147 86
GenBank mRNA AJ544829.2 - 10 - 11 1420 86

View Full Product Details