Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc37a4b
Entrez Gene ID
393914
Gene Name
solute carrier family 37 member 4b
Gene Aliases
G6pt1, slc37a4, zgc:55871
Chromosome Location
Chr.10: 39136106 - 39154626 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328095.1 NP_001315024.1 7 - 8 1758 77
XM_005169205.4 XP_005169262.1 7 - 8 1304 77
XM_009305596.3 XP_009303871.1 7 - 8 1336 77
XM_021479139.1 XP_021334814.1 7 - 8 1428 77

View Full Product Details