Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
txnl1
Entrez Gene ID
394113
Gene Name
thioredoxin-like 1
Gene Aliases
TXNL, zgc:55368
Chromosome Location
Chr.21: 918297 - 929448 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201138.2 NP_957432.2 2 - 3 353 74
XM_021468785.1 XP_021324460.1 2 - 3 327 74
GenBank mRNA BC045322.1 - 2 - 3 353 74
BC164656.1 - 2 - 3 231 74

View Full Product Details