Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
txnl1
Entrez Gene ID
394113
Gene Name
thioredoxin-like 1
Gene Aliases
TXNL, zgc:55368
Chromosome Location
Chr.21: 918297 - 929448 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201138.2 NP_957432.2 4 - 5 645 92
GenBank mRNA BC045322.1 - 4 - 5 645 92
BC164656.1 - 4 - 5 523 92

View Full Product Details