Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mapk3
Entrez Gene ID
399480
Gene Name
mitogen-activated protein kinase 3
Gene Aliases
ERK1, fi06b09, wu:fi06b09, zERK1
Chromosome Location
Chr.3: 26211974 - 26222308 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201507.1 NP_958915.1 8 - 9 1351 67
GenBank mRNA AB030902.1 - 8 - 9 1270 67
BC045505.1 - 8 - 9 1351 67
BC066401.1 - 8 - 9 1339 67
BC097073.1 - 8 - 9 1424 67

View Full Product Details