Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
snx16
Entrez Gene ID
402964
Gene Name
sorting nexin 16
Gene Aliases
zgc:77396
Chromosome Location
Chr.24: 42115501 - 42121814 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205665.1 NP_991228.1 7 - 7 850 71
GenBank mRNA BC065659.1 - 7 - 7 850 71

View Full Product Details