Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
grnb
Entrez Gene ID
335453
Gene Name
granulin b
Gene Aliases
fj14b08, wu:fj14b08, zgc:111858
Chromosome Location
Chr.24: 36339224 - 36364786 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212738.1 NP_997903.1 8 - 9 782 61
XM_021470114.1 XP_021325789.1 8 - 9 1105 61
GenBank mRNA AY289606.1 - 8 - 9 782 61
BC096854.1 - 8 - 9 776 61
BC163965.1 - 6 - 7 689 61

View Full Product Details